4-Amino-5-fluoro-1-((2R,5S)-rel-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one

Lieferant: Apollo Scientific

PC400599-100MG
APOSPC400599-100MGEA 274 EUR
APOSPC400599-100MG
4-Amino-5-fluoro-1-((2R,5S)-rel-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one
4-Amino-5-fluoro-1-((2R,5S)-rel-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one
Formel: C₈H₁₀FN₃O₃S
Molecular Weight: 247,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00870151
CAS-Nummer: 143491-54-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR