(3-Chloro-5-(trifluoromethyl)pyridin-2-yloxy)acetic acid
Lieferant: Apollo Scientific
PC400633-1G
PC400633-250MG
PC400633-500MG
PC400633-2.5G
APOSPC400633-1GEA
0
EUR
APOSPC400633-1G
APOSPC400633-250MG
APOSPC400633-500MG
APOSPC400633-2.5G
(3-Chloro-5-(trifluoromethyl)pyridin-2-yloxy)acetic acid
(3-Chloro-5-(trifluoromethyl)pyridin-2-yloxy)acetic acid
Formel:
C₈H₅ClF₃NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD10568276 CAS-Nummer: 95355-49-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...