4-(4-Trifluoromethylpyridin-2-yl)benzoic acid

Lieferant: Apollo Scientific

PC400664-1G PC400664-250MG PC400664-500MG
APOSPC400664-1GEA 0 EUR
APOSPC400664-1G APOSPC400664-250MG APOSPC400664-500MG
4-(4-Trifluoromethylpyridin-2-yl)benzoic acid
4-(4-Trifluoromethylpyridin-2-yl)benzoic acid
Formel: C₁₃H₈F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18384864
CAS-Nummer: 1299607-71-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR