5-Bromo-2-(4-methylpiperazin-1-yl)-3-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-2-(4-methylpiperazino)-3-(trifluoromethyl)pyridine

PC400689-1G PC400689-250MG PC400689-500MG PC400689-2.5G
APOSPC400689-1GEA 387 EUR
APOSPC400689-1G APOSPC400689-250MG APOSPC400689-500MG APOSPC400689-2.5G
5-Bromo-2-(4-methylpiperazin-1-yl)-3-(trifluoromethyl)pyridine
5-Bromo-2-(4-methylpiperazin-1-yl)-3-(trifluoromethyl)pyridine
Formel: C₁₁H₁₃BrF₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD15142779
CAS-Nummer: 1208081-84-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR