2-(3,4-Difluorophenyl)ethanol 97%
Lieferant: Apollo Scientific
PC400701-5G
APOSPC400701-5GEA
422
EUR
APOSPC400701-5G
2-(3,4-Difluorophenyl)ethanol 97%
2-(3,4-Difluorophenyl)ethanol
Formel:
C₈H₈F₂O Siedepunkt: 69 °C/0,1mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143061 CAS-Nummer: 286440-92-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...