2-(Trifluoromethyl)pyrimidin-5-ol 97%

Lieferant: Apollo Scientific
Achtung

PC400703-100MG
APOSPC400703-100MGEA 81.2 EUR
APOSPC400703-100MG
2-(Trifluoromethyl)pyrimidin-5-ol 97%
2-(Trifluoromethyl)pyrimidin-5-ol
Formel: C₅H₃F₃N₂O
Molecular Weight: 164,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10696944
CAS-Nummer: 100991-09-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR