3-Fluor-5-hydroxybenzonitril 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC400705-25G
APOSPC400705-25GEA
119
EUR
APOSPC400705-25G
3-Fluor-5-hydroxybenzonitril 97%
3-Fluor-5-hydroxybenzonitril
Formel:
C₇H₄FNO Molecular Weight: 137.1112 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09972133 CAS-Nummer: 473923-95-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...