3-(2,3-Difluorophenyl)propanoic acid

Lieferant: Apollo Scientific
Achtung

PC400709-25G PC400709-5G
APOSPC400709-25GEA 316 EUR
APOSPC400709-25G APOSPC400709-5G
3-(2,3-Difluorophenyl)propanoic acid
3-(2,3-Difluorophenyl)propanoic acid
Formel: C₉H₈F₂O₂
Molecular Weight: 186.155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04116051
CAS-Nummer: 412961-26-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR