5-Chloro-6-fluoropyridin-3-ol

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-2-fluoro-5-hydroxypyridine

PC400723-1G
APOSPC400723-1GEA 194 EUR
APOSPC400723-1G
5-Chloro-6-fluoropyridin-3-ol
5-Chloro-6-fluoropyridin-3-ol
Formel: C5H3ClFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD08061311
CAS-Nummer: 209328-72-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR