5-Bromo-3-chloro-2-fluoropyridine 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC400730-25G
APOSPC400730-25GEA
85.7
EUR
APOSPC400730-25G
5-Bromo-3-chloro-2-fluoropyridine 98%
5-Bromo-3-chloro-2-fluoropyridine
Formel:
C₅H₂BrClFN Molecular Weight: 210.43 g/mol Schmelzpunkt: 60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09834365 CAS-Nummer: 38185-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...