2-Hydroxy-5-(trifluormethyl)benzonitril

Lieferant: Apollo Scientific

Synonyme: 2-Hydroxy-5-trifluoromethylbenzonitrile

PC400733-5G
APOSPC400733-5GEA 172 EUR
APOSPC400733-5G
2-Hydroxy-5-(trifluormethyl)benzonitril
2-Hydroxy-5-(trifluormethyl)benzonitril
Formel: C₈H₄F₃NO
Molecular Weight: 187,1187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11213679
CAS-Nummer: 142167-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR