5-Fluoropyrimidin-2-amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-5-fluoropyrimidine
PC400734-5G
APOSPC400734-5GEA
133
EUR
APOSPC400734-5G
5-Fluoropyrimidin-2-amine
5-Fluoropyrimidin-2-amine
Formel:
C4H4FN3 Molecular Weight: 113,0931 g/mol Schmelzpunkt: 192…193 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11846500 CAS-Nummer: 1683-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...