4-Bromo-6-(trifluoromethyl)pyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
PC400735-5G
APOSPC400735-5GEA
179
EUR
APOSPC400735-5G
4-Bromo-6-(trifluoromethyl)pyrimidine
4-Bromo-6-(trifluoromethyl)pyrimidine
Formel:
C₅H₂BrF₃N₂ Molecular Weight: 226.9800 g/mol Siedepunkt: 36…37 °C Schmelzpunkt: 13…15 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10574962 CAS-Nummer: 785777-89-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...