2,5-Difluor-p-toluylsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,5-Difluoro-4-methylbenzoic acid

PC400739-5G
APOSPC400739-5GEA 65.4 EUR
APOSPC400739-5G
2,5-Difluor-p-toluylsäure
2,5-Difluor-p-toluylsäure
Formel: C₈H₆F₂O₂
Molecular Weight: 172,13 g/mol
Schmelzpunkt: 169…171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08063230
CAS-Nummer: 103877-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR