3-(1H,1H-Nonafluoropentyl)-4-methylcyclopentane-1,1-dicarboxylic acid

Lieferant: Apollo Scientific
Achtung

PC4009-250MG
APOSPC4009-250MGEA 224 EUR
APOSPC4009-250MG
3-(1H,1H-Nonafluoropentyl)-4-methylcyclopentane-1,1-dicarboxylic acid
3-(1H,1H-Nonafluoropentyl)-4-methylcyclopentane-1,1-dicarboxylic acid
Formel: C₁₃H₁₃F₉O₄
Schmelzpunkt: 130…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01320755
CAS-Nummer: 20116-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR