Methyl (3-chloro-5-(trifluoromethyl)pyridin-2-ylsulphanyl)acetate

Lieferant: Apollo Scientific
Achtung

PC401003-1G
APOSPC401003-1GEA 121 EUR
APOSPC401003-1G
Methyl (3-chloro-5-(trifluoromethyl)pyridin-2-ylsulphanyl)acetate
Methyl (3-chloro-5-(trifluoromethyl)pyridin-2-ylsulphanyl)acetate
Formel: C₉H₇ClF₃NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00245199
CAS-Nummer: 1024368-23-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR