Ethyl-2-(trifluoromethyl)thiazole-4-carboxylate

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-(trifluoromethyl)thiazole-4-carboxylate

PC401006-1G
APOSPC401006-1GEA 132 EUR
APOSPC401006-1G
Ethyl-2-(trifluoromethyl)thiazole-4-carboxylate
Ethyl-2-(trifluoromethyl)thiazole-4-carboxylate
Formel: C₇H₆F₃NO₂S
Molecular Weight: 225,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10568298
CAS-Nummer: 133046-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR