1-tert-Butoxycarbonyl-4-(4-fluorobenzoyl)piperidine 96%

Lieferant: Apollo Scientific

Synonyme: 4-(4-Fluorobenzoyl)piperidine-1-carboxylic acid tert-butyl ester, tert-Butyl 4-(4-fluorobenzoyl)piperidine-1-carboxylate

PC401007-1G
APOSPC401007-1GEA 192 EUR
APOSPC401007-1G
1-tert-Butoxycarbonyl-4-(4-fluorobenzoyl)piperidine 96%
1-tert-Butoxycarbonyl-4-(4-fluorobenzoyl)piperidine
Formel: C₁₇H₂₂FNO₃
Molecular Weight: 307,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114990
CAS-Nummer: 160296-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR