5-(Trifluoromethy)pyridine-2-carboxamidine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC401009-1G
APOSPC401009-1GEA 85.7 EUR
APOSPC401009-1G
5-(Trifluoromethy)pyridine-2-carboxamidine hydrochloride
5-(Trifluoromethy)pyridine-2-carboxamidine hydrochloride
Formel: C₇H₇ClF₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00203995
CAS-Nummer: 175277-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR