5-[2-(Trifluoromethyl)phenyl]cyclohexane-1,3-dione

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Hydroxy-5-[2-(trifluoromethyl)phenyl]cyclohex-2-ene-1-one

PC401019-1G
APOSPC401019-1GEA 366 EUR
APOSPC401019-1G
5-[2-(Trifluoromethyl)phenyl]cyclohexane-1,3-dione
5-[2-(Trifluoromethyl)phenyl]cyclohexane-1,3-dione
Formel: C₁₃H₁₁F₃O₂
Molecular Weight: 256,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03844403
CAS-Nummer: 55579-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR