2-Cyano-3-(methylamino)-5-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific

PC401037-1G
APOSPC401037-1GEA 364 EUR
APOSPC401037-1G
2-Cyano-3-(methylamino)-5-(trifluoromethyl)pyridine
2-Cyano-3-(methylamino)-5-(trifluoromethyl)pyridine
Formel: C₈H₆F₃N₃
Molecular Weight: 201,149 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25970568
CAS-Nummer: 1449117-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR