3-[Bis(2-hydroxyethyl)amino]-2-cyano-5-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific

PC401038-1G
APOSPC401038-1GEA 783 EUR
APOSPC401038-1G
3-[Bis(2-hydroxyethyl)amino]-2-cyano-5-(trifluoromethyl)pyridine
3-[Bis(2-hydroxyethyl)amino]-2-cyano-5-(trifluoromethyl)pyridine
Formel: C₁₁H₁₂F₃N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28122855
CAS-Nummer: 1672655-86-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR