Ethyl 2-[[2-cyano-5-(trifluoromethyl)pyridin-3-yl]amino]acetate

Lieferant: Apollo Scientific

PC401039-1G
APOSPC401039-1GEA 364 EUR
APOSPC401039-1G
Ethyl 2-[[2-cyano-5-(trifluoromethyl)pyridin-3-yl]amino]acetate
Ethyl 2-[[2-cyano-5-(trifluoromethyl)pyridin-3-yl]amino]acetate
Formel: C₁₁H₁₀F₃N₃O₂
Molecular Weight: 273,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25970571
CAS-Nummer: 1449117-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR