3-(Dimethylamino)-2-cyano-5-(trifluoromethyl)pyridine
Lieferant: Apollo Scientific
PC401040-1G
APOSPC401040-1GEA
783
EUR
APOSPC401040-1G
3-(Dimethylamino)-2-cyano-5-(trifluoromethyl)pyridine
3-(Dimethylamino)-2-cyano-5-(trifluoromethyl)pyridine
Formel:
C₉H₈F₃N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00205839 CAS-Nummer: 257862-49-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...