2-Cyano-3-phenylsulphanyl-5-(trifluoromethyl)pyridine
Lieferant: Apollo Scientific
PC401043-1G
APOSPC401043-1GEA
371
EUR
APOSPC401043-1G
2-Cyano-3-phenylsulphanyl-5-(trifluoromethyl)pyridine
Formel:
C₁₃H₇F₃N₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD25970575 CAS-Nummer: 1449117-61-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...