2-Cyano-5-(trifluoromethyl)-3-[4-(trifluoromethyl)phenoxy]pyridine
Lieferant: Apollo Scientific
PC401045-1G
APOSPC401045-1GEA
364
EUR
APOSPC401045-1G
2-Cyano-5-(trifluoromethyl)-3-[4-(trifluoromethyl)phenoxy]pyridine
2-Cyano-5-(trifluoromethyl)-3-[4-(trifluoromethyl)phenoxy]pyridine
Formel:
C₁₄H₆F₆N₂O Molecular Weight: 332,201 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00664116 CAS-Nummer: 338759-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...