3-Chloro-1-methyl-5-(trifluoromethyl)pyridin-2-one 95%

Lieferant: Apollo Scientific
Achtung

PC401046-1G PC401046-250MG PC401046-5G
APOSPC401046-1GEA 447 EUR
APOSPC401046-1G APOSPC401046-250MG APOSPC401046-5G
3-Chloro-1-methyl-5-(trifluoromethyl)pyridin-2-one 95%
3-Chloro-1-methyl-5-(trifluoromethyl)pyridin-2-one
Formel: C₇H₅ClF₃NO
Molecular Weight: 211.569 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28122856
CAS-Nummer: 1281189-85-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR