3-Chloro-5-(trifluoromethyl)pyridine-2-carboxamidine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-5-(trifluoromethyl)picolinimidamide hydrochloride

PC401061-1G
APOSPC401061-1GEA 183 EUR
APOSPC401061-1G
3-Chloro-5-(trifluoromethyl)pyridine-2-carboxamidine hydrochloride
3-Chloro-5-(trifluoromethyl)pyridine-2-carboxamidine hydrochloride
Formel: C₇H₅ClF₃N₃·ClH
Molecular Weight: 260,044 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11975591
CAS-Nummer: 1179362-45-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR