3-Bromo-1-methyl-5-(trifluoromethyl)pyridin-2-one

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-1-methyl-5-(trifluoromethyl)pyridin-2(1H)-one

PC401073-1G
APOSPC401073-1GEA 139 EUR
APOSPC401073-1G
3-Bromo-1-methyl-5-(trifluoromethyl)pyridin-2-one
3-Bromo-1-methyl-5-(trifluoromethyl)pyridin-2-one
Formel: C₇H₅BrF₃NO
Molecular Weight: 256,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15143623
CAS-Nummer: 1215205-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR