6-Fluoropyridine-3,4-diamine

Lieferant: Apollo Scientific

PC401502-1G
APOSPC401502-1GEA 727 EUR
APOSPC401502-1G
6-Fluoropyridine-3,4-diamine
6-Fluoropyridine-3,4-diamine
Formel: C₅H₆FN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09743966
CAS-Nummer: 60186-24-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR