4,4-Difluoropiperidine 97%

Lieferant: Apollo Scientific
Achtung

PC401503-1G
APOSPC401503-1GEA 21.9 EUR
APOSPC401503-1G
4,4-Difluoropiperidine 97%
4,4-Difluoropiperidine
Formel: C₅H₉F₂N
Molecular Weight: 121,128 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03094281
CAS-Nummer: 21987-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR