4-[(Trifluoromethyl)thio]benzamide

Lieferant: Apollo Scientific
Achtung

PC4019-1G
APOSPC4019-1GEA 37.7 EUR
APOSPC4019-1G
4-[(Trifluoromethyl)thio]benzamide
4-[(Trifluoromethyl)thio]benzamide
Formel: C₈H₆F₃NOS
Molecular Weight: 221,2 g/mol
Schmelzpunkt: 126…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02689728
CAS-Nummer: 330-15-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR