tert-Butyl (3-amino-2,6-difluorophenyl)carbamate
Lieferant: Apollo Scientific
PC402000-250MG
APOSPC402000-250MGEA
31.9
EUR
APOSPC402000-250MG
tert-Butyl (3-amino-2,6-difluorophenyl)carbamate
tert-Butyl (3-amino-2,6-difluorophenyl)carbamate
Formel:
C₁₁H₁₄F₂N₂O₂ Molecular Weight: 244,238 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13182479 CAS-Nummer: 535170-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...