tert-Butyl 3-(bromomethyl)-5-fluoro-1H-indazole-1-carboxylate

Lieferant: Apollo Scientific

PC402003-250MG
APOSPC402003-250MGEA 539 EUR
APOSPC402003-250MG
tert-Butyl 3-(bromomethyl)-5-fluoro-1H-indazole-1-carboxylate
tert-Butyl 3-(bromomethyl)-5-fluoro-1H-indazole-1-carboxylate
Formel: C₁₃H₁₄BrFN₂O₂
Molecular Weight: 329,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10696845
CAS-Nummer: 944904-75-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR