7-Fluoro-1H-indazole-3-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
PC402004-250MG
APOSPC402004-250MGEA
864
EUR
APOSPC402004-250MG
7-Fluoro-1H-indazole-3-carbaldehyde
7-Fluoro-1H-indazole-3-carbaldehyde
Formel:
C₈H₅FN₂O Lagertemperatur: Kühlschrank |
MDL:
MFCD13176919 CAS-Nummer: 900506-29-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...