4-Fluoro-3aH-indazol-3-amine

Lieferant: Apollo Scientific

PC402006-250MG
APOSPC402006-250MGEA 510 EUR
APOSPC402006-250MG
4-Fluoro-3aH-indazol-3-amine
4-Fluoro-3aH-indazol-3-amine
Formel: C₇H₆FN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05855527
CAS-Nummer: 697230-91-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR