4-Fluoro-3-methyl-1H-indazole

Lieferant: Apollo Scientific

PC402007-250MG
APOSPC402007-250MGEA 655 EUR
APOSPC402007-250MG
4-Fluoro-3-methyl-1H-indazole
4-Fluoro-3-methyl-1H-indazole
Formel: C₈H₇FN₂
Molecular Weight: 150,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04109834
CAS-Nummer: 662146-05-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR