7-Bromo-5-fluoro-1H-indazole

Lieferant: Apollo Scientific

PC402013-250MG
APOSPC402013-250MGEA 655 EUR
APOSPC402013-250MG
7-Bromo-5-fluoro-1H-indazole
7-Bromo-5-fluoro-1H-indazole
Formel: C₇H₄BrFN₂
Molecular Weight: 215,022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21602945
CAS-Nummer: 1100214-35-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR