2-(4-Fluorophenyl)-1H-imidazole

Lieferant: Apollo Scientific

PC402015-1G PC402015-250MG
APOSPC402015-1GEA 154 EUR
APOSPC402015-1G APOSPC402015-250MG
2-(4-Fluorophenyl)-1H-imidazole
2-(4-Fluorophenyl)-1H-imidazole
Formel: C₉H₇FN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04114652
CAS-Nummer: 4278-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR