2,2,2-Trifluoro-1-(1H-indol-5-yl)ethanone
Lieferant: Apollo Scientific
PC402020-250MG
APOSPC402020-250MGEA
306
EUR
APOSPC402020-250MG
2,2,2-Trifluoro-1-(1H-indol-5-yl)ethanone
2,2,2-Trifluoro-1-(1H-indol-5-yl)ethanone
Formel:
C₁₀H₆F₃NO Molecular Weight: 213,156 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13173250 CAS-Nummer: 170366-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...