(4-Fluorophenyl)(1H-indol-7-yl)methanone

Lieferant: Apollo Scientific

PC402021-250MG
APOSPC402021-250MGEA 582 EUR
APOSPC402021-250MG
(4-Fluorophenyl)(1H-indol-7-yl)methanone
(4-Fluorophenyl)(1H-indol-7-yl)methanone
Formel: C₁₅H₁₀FNO
Molecular Weight: 239,244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13177625
CAS-Nummer: 405275-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR