7-(Trifluoromethoxy)-1H-indole
Lieferant: Apollo Scientific
PC402022-250MG
APOSPC402022-250MGEA
531
EUR
APOSPC402022-250MG
7-(Trifluoromethoxy)-1H-indole
7-(Trifluoromethoxy)-1H-indole
Formel:
C₉H₆F₃NO Molecular Weight: 201,145 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11109870 CAS-Nummer: 396075-91-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...