tert-Butyl 6-fluoro-2,3-dihydro-1H-indole-1-carboxylate

Lieferant: Apollo Scientific

PC402023-1G
APOSPC402023-1GEA 786 EUR
APOSPC402023-1G
tert-Butyl 6-fluoro-2,3-dihydro-1H-indole-1-carboxylate
tert-Butyl 6-fluoro-2,3-dihydro-1H-indole-1-carboxylate
Formel: C₁₃H₁₆FNO₂
Molecular Weight: 237,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13183504
CAS-Nummer: 1065183-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR