4-(Trifluoromethyl)indoline

Lieferant: Apollo Scientific

PC402024-250MG
APOSPC402024-250MGEA 191 EUR
APOSPC402024-250MG
4-(Trifluoromethyl)indoline
4-(Trifluoromethyl)indoline
Formel: C₉H₈F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD08544326
CAS-Nummer: 905274-07-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR