4-Fluoro-2,3-dihydro-1H-isoindol-1-one
Lieferant: Apollo Scientific
|
Achtung
|
PC402025-250MG
APOSPC402025-250MGEA
582
EUR
APOSPC402025-250MG
4-Fluoro-2,3-dihydro-1H-isoindol-1-one
4-Fluoro-2,3-dihydro-1H-isoindol-1-one
Formel:
C₈H₆FNO Molecular Weight: 151,138 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13177756 CAS-Nummer: 366452-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...