7-(4-Fluorophenyl)-2,3-dihydro-1H-isoindol-1-one
Lieferant: Apollo Scientific
PC402027-250MG
APOSPC402027-250MGEA
611
EUR
APOSPC402027-250MG
7-(4-Fluorophenyl)-2,3-dihydro-1H-isoindol-1-one
7-(4-Fluorophenyl)-2,3-dihydro-1H-isoindol-1-one
Formel:
C₁₄H₁₀FNO Molecular Weight: 227,234 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13177841 CAS-Nummer: 200049-49-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...