4-Bromo-5-fluorobenzene-1,2-diol

Lieferant: Apollo Scientific

PC402028-250MG
APOSPC402028-250MGEA 495 EUR
APOSPC402028-250MG
4-Bromo-5-fluorobenzene-1,2-diol
4-Bromo-5-fluorobenzene-1,2-diol
Formel: C₆H₄BrFO₂
Molecular Weight: 206,997 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13177846
CAS-Nummer: 656804-73-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR