3-Fluoro-4-(1H-pyrazolo[3,4-b]pyridin-4-yloxy)aniline

Lieferant: Apollo Scientific

PC402030-250MG
APOSPC402030-250MGEA 757 EUR
APOSPC402030-250MG
3-Fluoro-4-(1H-pyrazolo[3,4-b]pyridin-4-yloxy)aniline
3-Fluoro-4-(1H-pyrazolo[3,4-b]pyridin-4-yloxy)aniline
Formel: C₁₂H₉FN₄O
Molecular Weight: 244,224 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13178100
CAS-Nummer: 1115490-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR