4-Fluoropyridine-2-carbonitrile
Lieferant: Apollo Scientific
|
Gefahr
|
PC402032-1G
APOSPC402032-1GEA
364
EUR
APOSPC402032-1G
4-Fluoropyridine-2-carbonitrile
4-Fluoropyridine-2-carbonitrile
Formel:
C₆H₃FN₂ Schmelzpunkt: 72…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09834127 CAS-Nummer: 847225-56-3 UN: 3439 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...