4-Methyl-6-trifluoromethylpyridin-3-ylamine

Lieferant: Apollo Scientific

PC402033-250MG
APOSPC402033-250MGEA 539 EUR
APOSPC402033-250MG
4-Methyl-6-trifluoromethylpyridin-3-ylamine
4-Methyl-6-trifluoromethylpyridin-3-ylamine
Formel: C₇H₇F₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11848206
CAS-Nummer: 944317-54-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR